Learn More
Hanna Pharmaceutical Supply Co., Inc. Empty INTRAVIA Container 500mL
Supplier: Hanna Pharmaceutical Supply Co., Inc. 2J8003
Empty INTRAVIA Container 500 mL. Sterile, nonpyrogenic fluid path. Port Tubes, which contact fluid path are Non-DEHP PVC. Blue port protector, which does not contact fluid path, contains DEHP-plasticized PVC. 48/CS
Please note: This item is nonrefundable, non-returnable and non-cancelable once order is placed.
This item is not returnable.
View return policy