Learn More
World Precision Instrument Battery Operated Impedance Measurement
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: World Precision Instrument SYSOMEGAZ
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Measure mV and MΩ impedance of metal or glass microelectrodes
Omega-Tip-Z™ was created especially for measuring impedance in etched tungsten, platinum-iridium* and steel microelectrodes, as well as electrolyte-filled micropipettes. The meter’s AC impedance-measuring circuit is unaffected by electrode offset or tip junction potentials. The gold-plated miniature probe lets you conveniently monitor micro-electrode impedance in electrolytes, and an electrode tip cleaning feature lets you remove buildup quickly. Omega-Tip-Z can also measure DC electrode tip potentials up to 2000 millivolts. The instrument operates for hundreds of hours without battery failure.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.