Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Basic Fixed Magnification Stereo Microscope
Basic Fixed Magnification Stereo Microscope is ideal for simple jobs.
Description
Optical System
- 10X WF eyepiece
- Dual magnification
Viewing Head
- Binocular viewing head with 45° incline
- Interpupillary adjustment of 55mm to 75mm
- Diopter Adjustment
Focusing
- Coarse focus adjustment
- 40mm height adjustment range on focusing mechanism
- 70mm working distance
Stand
- Low profile plain stage with clips
- Solid stand, compact design
Illumination
- Eco-friendly and energy efficient LED illumination
- LED ring in transmitted light, provides homogeneous light distribution
- LED incident Illumination
- Membrane switch controls
- Corded; no battery option
Specifications
Corded; no battery | |
5 years optical and mechanical, 1 year electrical from date of purchase | |
WF 10X |
1X, 3X | |
Binocular, 45° inclined |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction