Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pyramex™ Baseball Bump Cap Head Protection
Baseball Bump Cap Head Protection offers a bump cap and baseball cap in one.
$176.00 - $203.00
Specifications
Certifications/Compliance | Non-rated |
---|---|
For Use With (Application) | Meter readers, food processors, and other industries that do not require full hard hat protection |
Material | Cotton, Polyester, and Inner shell of high impact acrylonitrile-butadiene-styrene |
Product Type | Bump Cap |
Size | 52 to 61 cm |
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
19-155-462
|
Pyramex™
HP50011 |
Black |
Case of 12 for $176.00
|
|
|||||
19-155-463
|
Pyramex™
HP50031 |
Lime Green |
Case of 12 for $203.00
|
|
|||||
Description
- Reflective piping adds extra visibility
- Soft inner foam cushion for comfortable wear
- Size can be adjusted with a hook and loop strap to give a quick, secure fit
- Tough polyethylene vented shell is removable and the outer cap is fully washable
- Ideal for food processing and other industries that don't require full hard hat protection
- Holes in the liner match up with two mesh panels on the cap to provide additional ventilation for coolness and comfort
Specifications
Non-rated | |
Cotton, Polyester, and Inner shell of high impact acrylonitrile-butadiene-styrene | |
52 to 61 cm | |
Not Vented |
Meter readers, food processors, and other industries that do not require full hard hat protection | |
Bump Cap | |
Baseball Bump Cap | |
Short Brim (Cap Style) |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title