Learn More
Hach Company BART Test Combination Package: Iron-Related Bacteria, Sulfate-Reducing Bacteria, Slime-Forming Bacteria (3 each)
Supplier: Hach Company 2434809
Combination pack: convenient multi-pack for detection (3 each of IRB/SRB/SLYM) of iron-related, sulfate-reducing, and slime-forming bacteria. BART biodetectors are excellent diagnostic tools to help identify the presence of process disruptors. Find out if you have bacteria creating slime layers and limiting disinfection, or discover if iron-reducing bacteria are eating your pipes.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.