Learn More
Accuform™ Barricade Light
Accuform™ Barricade Lights add visibility and draw attention to warn of hazards situations in low-light conditions
$75.50 - $126.50
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
19-169-600
|
Accuform™
FBL182 |
Incandescent Barricade Light |
Each for $75.50
|
|
|||||
19-169-601
|
Accuform™
FBL639 |
Each for $126.50
|
|
||||||
Description
Incandescent:
- Durable polycarbonate 7 in. amber lens with plastic yellow battery-case base
- Incandescent light can be set to steady-on or flashing
- Photocell turns on automatically in dark conditions
- Concealed on-off switch with bolt and set-in nut included for easy mounting
- Requires two, 6-volt batteries (not included)
Solar:
- Eliminate the maintenance, replacement, and disposal costs of batteries and bulbs
- Solar photovoltaic cells located on top of the 7 in. diameter x 3 ¾” high-impact, polycarbonate lens
- Durable polypropylene case has universal mount bracket and 360° swivel
- On/off and steady-on/flash switches secured inside small holes
- High-intensity LED with steady-on brightness of 2000 MCD, or flashing brightness of 4000 MCD with flash rate of 60 per minute
- Batteries operate up to three nights on single charge
- Photocell activates device when ambient light falls below 215 lux
Specifications
Amber-Yellow | |
Polycarbonate |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.