Learn More
Cole-Parmer Oakton Wd-03316-80:Barometer W Digital Thermometer
Supplier: Cole-Parmer 0331680
Oakton Barometer with Digital Thermometer measures atmospheric pressure and temperature. Barometer is encased in corrosion-resistant ABS plastic housing. Includes wall mounting ring and instructions. Operates on one AA battery (included). Measures 1.3"D x 5.3" dia.