Learn More
Cole-Parmer Cole-Parmer Temperature Compensated Barometer, 930 to 1070 mbar, 27.5 to 31.6" Hg
Supplier: Cole-Parmer 0331670
Cole-Parmer Temperature Compensated Barometers measure atmospheric pressure in millibar (mbar) and inches Hg or millibar and mm Hg. Barometers also monitor temperature and provide temperature compensation with a built-in bi-metal thermometer. Ideal for monitoring impending weather changes. Barometers are encased in brass, with bezel for bulkhead mounting. Measure 2"D x 6.1" dia.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.