Learn More
Thermo Scientific™ Barnstead™ Easypure™ Cartridges and Filters
Extend the life of your Thermo Scientific™ Barnstead™ Easypure™ II or Easypure™ RF water system with replacement cartridges and filters.
Supplier: Thermo Scientific™ D502127
Description
Extend the life of your Thermo Scientific™ Barnstead™ Easypure™ II or Easypure™ RF water system with Barnstead™ Easypure™ Cartridges and Filters. Choose the cartridge pack according to your feed water and system.
D502126:
- Includes: Organic/Colloid; two Thermo Scientific™ Ultrapure™ Ion Exchange
- Type 1 filter
D502124:
- Includes: Organic/Colloid, Ultrapure Ion Exchange, Organic Free Organic Removal
- Ultralow filter with Organic Free Cartridge
D502127:
- Includes: 09-034-18 Organic/Ion Exchange, two 09-050-46 Ultrapure Ion Exchange
- Type 1 filter
D502125:
- Includes: 09-034-18 Organic/Ion Exchange, 09-050-46 Ultrapure Ion Exchange, 09-034-16 Organic Free Organic Removal
- Ultralow filter with Organic Free Cartridge
Specifications
Cartridge pack; RO or distilled feed | |
Reverse Osmosis, Distilled | |
Easypure™ II and Easypure UF | |
Cartridge Pack |
missing translation for 'provideContentCorrection'
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
missing translation for 'productTitle'
Spot an opportunity for improvement?
missing translation for 'provideContentCorrection'