Learn More
Thermo Scientific™ Barnstead™ Easypure™ Cartridges and Filters
Extend the life of your Thermo Scientific™ Barnstead™ Easypure™ II or Easypure™ RF water system with replacement cartridges and filters.
Supplier: Thermo Scientific™ D502124
Description
Extend the life of your Thermo Scientific™ Barnstead™ Easypure™ II or Easypure™ RF water system with Barnstead™ Easypure™ Cartridges and Filters. Choose the cartridge pack according to your feed water and system.
D502126:
- Includes: Organic/Colloid; two Thermo Scientific™ Ultrapure™ Ion Exchange
- Type 1 filter
D502124:
- Includes: Organic/Colloid, Ultrapure Ion Exchange, Organic Free Organic Removal
- Ultralow filter with Organic Free Cartridge
D502127:
- Includes: 09-034-18 Organic/Ion Exchange, two 09-050-46 Ultrapure Ion Exchange
- Type 1 filter
D502125:
- Includes: 09-034-18 Organic/Ion Exchange, 09-050-46 Ultrapure Ion Exchange, 09-034-16 Organic Free Organic Removal
- Ultralow filter with Organic Free Cartridge
Specifications
Cartridge pack; deionized feed | |
Deionized | |
Easypure™ II UV and Easypure II UV/UF | |
Cartridge Pack |