Learn More
Fisherbrand™ Barcode Reader for Urine Analyzer and Urinalysis Reagent Strips
Optional Barcode Reader records patient ID, and verifies strip canister barcodes. It is for use with the Fisherbrand Urine Analyzer and the full range of Urine Reagent Strips. A substantial equivalent to the leading national brand.
Description
A CLIA Waived, easy-to-use system that provides fast, highly-accurate results for the general evaluation of health, aiding in the diagnosis and monitoring of metabolic or systemic diseases that affect kidney function, endocrine disorders and diseases or disorders of the urinary tract.
High Accuracy and Reliability:
- Automatic calibration before each test
- Up to 120 tests per hour in Continuous Mode
- Reads strips with up to 10 parameters, including
- Microalbumin/Creatinine
- Minimal training required
- Robust construction
- Data Management ready
- Includes RS232C and USB ports for data transfer to an external computer or LIS
- 510(k) Cleared and CLIA Waived
Specifications
RS-232, USB | |
Barcode Reader | |
Standard RS232C Port for Barcode Reader or Data Transfer, USB Port for Data Transfer, 25 Pin Parallel Port for External Printer |
None Included | |
Canister Codes and Patient ID |