Learn More
Sigma Aldrich Fine Chemicals Biosciences Barbituric acid, 67-52-7, MFCD00006666, 100 g
Supplier: Sigma Aldrich Fine Chemicals Biosciences 185698100G
Empirical Formula (Hill Notation): C4H4N2O3, Molecular Weight: 128.09, ReagentPlus, 99%, mp: 248-252 °C, Synonym: 2,4,6-Trihydroxypyrimidine, Malonylurea.