Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ BAM 15
Mitochondrial protonophore uncoupler
$261.50 - $1066.00
Specifications
Chemical Name or Material | N5,N6-bis(2-Fluorophenyl)-[1,2,5]oxadiazolo[3,4-b]pyrazine-5,6-diamine |
---|---|
CAS | 210302-17-3 |
Target | Oxidative Phosphorylation Inhibitors |
Molecular Formula | C16H10F2N6O |
Formula Weight | 340.29 |
Description
Mitochondrial protonophore uncoupler; uncouples oxidative phosphorylation in L6 myoblast mitochondria in vitro (EC50 = 270 nM). Does not depolarize the plasma membrane. Protects mice from acute renal ischemic-reperfusion injury.Specifications
N5,N6-bis(2-Fluorophenyl)-[1,2,5]oxadiazolo[3,4-b]pyrazine-5,6-diamine | |
Oxidative Phosphorylation Inhibitors | |
340.29 |
210302-17-3 | |
C16H10F2N6O | |
>98% |
Safety and Handling
Recommended Storage : Store at -20°C
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title