Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher Science Education™ Balloons for Science Kits

Consumbles for use with multiple projects and science kits.
This item is not returnable.
View return policy
Description
- Round
- Oblong
Specifications
Balloon | |
For use in multiple projects and in science kits | |
9 in. |
Dark Blue | |
Round | |
23 cm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title