Learn More
iQ Biosciences BALB/c Mouse Bone Marrow Cells, 20M cells/vial
Supplier: iQ Biosciences IQBMBM102
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
High-quality, high-viability (typically > 70%) bone marrow cells from the BALB/c mouse, a commonly-used inbred mouse strain. Used for a wide variety of immunology-based applications, including cell population characterization and differentiation into specific cell types. All orders come with an iQ Certificate of Analysis.