Learn More
Andwin Scientific Vented Balance Enclosure 115V 36" Wide
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Andwin Scientific VBE306
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
![Request Bulk Order](/content/dam/fishersci/glyphs/15395_Quote_Bulk_EN.jpg)
Self-contained laboratory unit ideal for fine powders, chemicals and biological products. Designed with top-mounted HEPA/blower filtration set up to circualte internal chamber atmosphere, through the HEPA filter and into the lab. Constructed with optically clear 3/8" jeweled acrylic plastics and phenolic base with superior chemical resistance. Outside width dimension is 36". 115V.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.