Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
American Educational Products Bagged Specimen Collections
Each set contains six representative specimens.
Supplier: American Educational Products 2231B
Alert:
CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.
Includes: Identification key
Description
- Specimen size: 1.5 × 1.5 in. (3.8 × 3.8cm)
- Specimen names listed on bag
Specifications
Sedimentary Rock | |
Resealable zip lock bag with a label | |
6 Specimens |
5 to 12 | |
Specimen collections in bags | |
3 to 3.8 cm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction