Learn More
Interscience Lab Inc BAG RACK 80/100ML
Supplier: Interscience Lab Inc 221010
This item has a minimum qty of 3 per supplier requirements.
The BagRack bag holder allows for clean and methodical storage of bags (closed for example by BagClip). BagRack is made of stainless steel and is stackable and autoclavable. A drip tray is also available. 12.5 x 27.5 x 14 cm