Learn More
Uline Tyvek Bag Clay Desiccants, 1 x 2.5 x 0.25"""", 5 Gallon Pail
Supplier: Uline S5163
Clay desiccant in perforated sachet absorbs moisture from air when placed inside sealed container, adsorbing water vapor and maintaining low humidity to prevent spoilage and damage from excess moisture due to its porous clay with large surface area; allows air exchange while retaining beads.