Learn More
Chemglass Life Sciences PTFE Jacketed Stainless Steel Baffles Only: Overall Baffle Length: 360mm, Effective Baffle Length: 170mm, Baffle Width x Depth (mm): 7 x 10, PTFE Compression Fitting: 1/4", Fits Reactor Size: 300mL-500mL
Supplier: Chemglass Life Sciences CG1971M09
PTFE Jacketed Stainless Steel Baffles Only: Overall Baffle Length: 360mm, Effective Baffle Length: 170mm, Baffle Width x Depth (mm): 7 x 10, PTFE Compression Fitting: 1/4", Fits Reactor Size: 300mL-500mL. Designed to increase agitation in the reactor and mimic baffles found in larger glass lined reactors. PTFE jacketed Stainless Steel provides excellent strength and chemical durability. Pictured is a fully assembled baffle system, but this item is BAFFLE ONLY.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.