Learn More
Thermo Scientific™ BactiSwab™ Gel Collection and Transport Swabs: Dual Swab Amies Clear
Thermo Scientific™ Remel BactiSwab Gel Amies Clear are supplied with double plastic shaft.
Specifications
Media Type | Amies Clear |
---|---|
Closure Color | Blue |
Description | Amies Clear |
Quantity | 1 Tube/Envelope |
Shaft Material | Plastic |
Catalog Number | Mfr. No. | Packaging | SwabConfiguration | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Packaging | SwabConfiguration | Price | Quantity & Availability | ||||
R723005
|
Thermo Scientific™
R723005 |
50/Pk. | Single Applicator |
Pack of 50 for $99.00
|
N/A | ||||
R723000
|
Thermo Scientific™
R723000 |
500/Pk. | Single Applicator |
Case of 500 for $919.00
|
N/A | ||||
R723010
|
Thermo Scientific™
R723010 |
500/Pk. | Dual Applicator |
Case of 500 for $1,079.00
|
N/A | ||||
R723015
|
Thermo Scientific™
R723015 |
50/Pk. | Dual Applicator |
Pack of 50 for $118.00
|
N/A | ||||
Description
Thermo Scientific™ Remel BactiSwab Gel Amies Clear are supplied with double plastic shaft.
Rayon-tipped, plastic-shaft swab(s) are mounted in the cap of a translucent plastic tube with spiral chamber containing gel transport medium with or without charcoal.- Blue cap
- Round-bottom tube for easy racking and seamless molding to ensure strength and integrity
- Exclusive zip-seal packaging to promote optimum media performance
- Color-coded cap with protective overskirt
- Inert, non-toxic rayon swab tip for good sample retrieval and absorption
Specifications
Amies Clear | |
Amies Clear | |
Plastic |
Blue | |
1 Tube/Envelope |