Learn More
Electron Microscopy Sciences Bacti-Cinerator IV, 115 V
Supplier: Electron Microscopy Sciences 6242010
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The Bacti-Cinerator IV sterilizes by infrared heat (Heater Element contains no asbestos) and prevents infectious spatter and/or cross contamination. Organic material is incinerated deep inside the ceramic tunnel. To use, simply insert loop or needle into the tunnel. Complete sterilization takes only 5 to 7 seconds at optimum sterilizing temperature of 1,500 degree F. There is no need to wait for the loops or needles to turn red.
Unit does not require oxygen so it can be used in anaerobic chambers. Safeguard laboratory personnel with no open flame and no hazardous gas. UL approved.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.