Learn More
Sklar Instruments Backhaus Towel Clamp, 5-1/4 in., Curved, Perforating 1x1 Prongs, Pointed, EA.
Supplier: Sklar Instruments 472955
Backhaus Towel Clamp, 5-1/4 in., Curved, Perforating 1x1 Prongs, Pointed, EA. Sklar's Backhaus Towel Clamp is a perforating clamp used for grasping tissue, securing towels or drapes, and holding or reducing small bone fractures. This product is curved with 1X1 perforating prongs and 5-1/4 inches in length.