Learn More
Motic BA310E Microscope and Camera Bundle
The BA310 is designed for the rigors of daily routine work in the demanding applications of universities, clinics, laboratories, and many other life sciences or medical applications requiring quality optical performance.
Supplier: Motic 1100402700081
This item is not returnable.
View return policy
Description
- These models introduce a new rackless stage concept without a prominent gear rack to allow for a more convenient use of the X/Y movement.
- Allow easy interchangeability of its halogen bulb with LED modules
- Swiveling, 30° inclined Siedentopf type
- Reversed quintuple revolving nosepiece
- Coaxial coarse and fine focusing adjustment with tension adjustment
- Built in low position coaxial rackless stage (Right-hand control)
- Diopter Adjustment: ±5
Specifications
10X | |
BA310E LED Trinocular with BMH4000 Camera bundle | |
Moticam BMH4000 | |
WF 10X/20 mm |
CCIS™ EC Plan Achromat Objectives EC PL (4X, 10X, 40X(S), 100X(S-oil)) | |
Trinocular | |
LED |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov