Learn More
Jackson Immuno Research Labs Bovine Serum Albumin (IgG-Free, Protease-Free) 50.0 g
SDP
Supplier:  Jackson Immuno Research Labs 001000162
Encompass_Preferred
Bovine Serum Albumin (BSA) is used extensively as a carrier protein to dilute antibodies and as a general blocking agent in immunoassays and immunodetection protocols. Physical State: Freeze-dried solid Storage: Store freeze-dried powder at 2-8°C. Weigh out desired amount and dissolve in desired buffer. Prepare working dilution fresh each day.
Catalog No. NC9871802