Learn More
Vidaro™ B-Fore™ Boot Covers
Anti-static cleanroom boots feature a rigid sole and convenient color-coded tabs for easy size identification.
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
19-999-792
|
Vidaro™
B31585/XS |
Rigid tan sole; White; X-Small | Rigid tan sole; White; X-Small | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
19-999-793
|
Vidaro™
B31585/S |
Rigid tan sole; White; Small | Rigid tan sole; White; Small | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
19-999-784
|
Vidaro™
B31584/M |
Chemstat 939 Sole; White; Medium | Chemstat 939 Sole; White; Medium | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
19-051-394D
|
Vidaro™
B31585SP2/L (S/3264) |
Hard Sole; White; Large | Hard Sole; White; Large | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
19-051-394E
|
Vidaro™
B31585SP2/XL (S/3264) |
Rigid tan sole; White; X-Large | Rigid tan sole; White; X-Large | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
19-999-797
|
Vidaro™
B31585/2XL |
Rigid tan sole; White; XX-Large | Rigid tan sole; White; XX-Large | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
Description
Assure contamination control and increase worker comfort with reusable boots made from B-Fore high-performance ESD cleanroom fabric. Designed for the microelectronics industry and any applications where products must be protected from static dissipative charges, airborne particles, and lint. With a super-dense weave of 100% microdenier polyester fibers (over 19,000 filaments per square inch) B-Fore meets the highest standard of particulate control.
- Microengineered carbon-fiber grid dissipates electrostatic charges and minimizes manufacturing defects
- 19 in. high (48cm)
- Closure is strap-and-buckle at instep with adjustable snaps on the top tab
- Two snaps on the tab, four snaps on the hem
- One snap on back of leg to attach to a coverall
- Available in white, blue and orange with soft, rigid or cupped Chemstat 939™ soles
Specifications
White |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.