Learn More
ECM Bioscience β-Catenin Tyr-142 & Tyr-654 Phosphorylation Immunocytochemistry Kit, ECM Biosciences
Supplier: ECM Bioscience CK7610
The β-Catenin phospho-regulation kit can be used for immunocytochemical co-localization of β-Catenin phosphorylation at Tyr-142 and Tyr-654 relative to total β-Catenin. The kit includes rabbit polyclonal and mouse monoclonal antibodies, along with goat-anti-rabbit conjugated to DyLight 594 and goat anti-mouse conjugated to DyLight 488 for dual labeling experiments. Applications: ICC, IHC; Species Reactivity: Hu, Rt, Ms, Ck
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.