Learn More
CPC Scientific azido-valine-azobenzene-PEG-Biotin 5MG
SDP
Supplier:  CPC Scientific LINK003A
Encompass
An unlabeled "light" azobenzene-based cleavable linker (click-chemistry-compatible) for quantitative chemical proteomics.ONE-LETTER SEQUENCE: azido-V-azobenzene-PEG-BiotinMOLECULAR FORMULA: C45H67N11O9SMOLECULAR WEIGHT:938.15STORAGE CONDITIONS: -20 5CRESEARCH AREA: AntimicrobialREFERENCES: Yu Qian et al., ChemBioChem, 14, 1410 ? 1414 (2013)
Catalog No. 50-210-9675