Learn More
CPC Scientific azido-valine(13C5, 15N)-azobenzene-PEG-Biotin 25MG
SDP
Supplier:  CPC Scientific LINK004B
Encompass
An isotopically tagged azobenzene-based cleavable linker (click-chemistry-compatible) for quantitative chemical proteomics.ONE-LETTER SEQUENCE: azido-V-azobenzene-PEG-BiotinMOLECULAR FORMULA: C4013C5H67N1015NO9SMOLECULAR WEIGHT:944.1STORAGE CONDITIONS: -20 5CRESEARCH AREA: AntimicrobialREFERENCES: Yu Qian et al., ChemBioChem, 14, 1410 ? 1414 (2013)
Catalog No. 50-210-9678