Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dynalon™ Azlon™ Polypropylene Carboy
SDPSCI_ED

Catalog Number S01989D
Click to view available options
Capacity (Metric):
10 L

For Use With: Storing and dispensing liquids in educational settings.; Autoclavable; Closure Material: Polypropylene; Color: White

 

  • Autoclavable polypropylene, heavy-duty construction
  • Stylish rounded octagonal shape for increased rigidity when full
  • Offset neck for easier filling and pouring
  • Integral, comfortable blow molded handles for ease of carrying, are end-sealed to prevent contamination
  • Heavy-duty closures
  • Molded graduations allow easy viewing of contents

 

Product Type Carboy
Capacity (English) 2.6 gal.
Closure Material Polypropylene
Capacity (Metric) 10 L
Material Polypropylene
For Use With (Application) Storing and dispensing liquids in educational settings.
Includes Blow molded handles
Neck Style Offset
Shape Rounded Octagonal
Autoclavable Autoclavable
Color White
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit