Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Azelaic Acid, MP Biomedicals
Click to view available options
100g
Specifications
CAS | 123-99-9 |
Molecular Formula | C9H16O4 |
Synonym | azelaic acid, finacea, anchoic acid, azelex, lepargylic acid, skinoren, 1,7-heptanedicarboxylic acid, heptanedicarboxylic acid, 1,9-nonanedioic acid, emerox 1110 |
InChI Key | BDJRBEYXGGNYIS-UHFFFAOYSA-N |
SMILES | C(CCCC(=O)O)CCCC(=O)O |
IUPAC Name | nonanedioic acid |
Molecular Weight (g/mol) | 188.223 |
PubChem CID | 2266 |
ChEBI | CHEBI:48131 |
Formula Weight | 188.22 |
Show More |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction