Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Axygen™ Automation Tips for STAR, STARlet,STARplus and Nimbus™ Conductive Black, Liquid Level Sensing
Compatible with original equipment manufacturers' tips
Supplier: Axygen™ HT1000CBKHTRS5
Description
- Tip format: 96 tip
- Tip volume range: 10 μL to 1,000 μL
- Tip material: Conductive black or clear polypropylene
- RNase-/DNase-/human gDNA-free
- Nonpyrogenic
- Available options:
- With or without aerosol-resistant filters
- Nonsterile or sterile - Compatible head: 8-channel, 96-probe
- Rack format: ANSI/SLAS
- Rack material: Polypropylene
Specifications
1000 μL | |
Sterile | |
DNase, RNase Free | |
Rack, 96 | |
Polypropylene |
Non-Filtered | |
Black | |
Hamilton™ MICROLAB™ Nimbus, Hamilton™ MICROLAB™ STAR, Hamilton™ MICROLAB™ STARlet, Hamilton™ MICROLAB™ STARplus | |
Hamilton™ MICROLAB™ STAR, Hamilton™ MICROLAB™ STARlet, Hamilton™ MICROLAB™ STARplus, Hamilton™ MICROLAB™ Nimbus | |
Round |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title