Learn More
Axygen™ Automation Tips for Beckman Coulter™, 30μL, Clear, Filtered, Sterile, Maxymum Recovery™
![GREENER_CHOICE](/content/dam/fishersci/glyphs/greener_choice.png)
For complete compatibility and consistent fit
Supplier: Axygen™ FXF384XL96LRS
Alert:
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit
Description
Axygen brand automation tips are engineered to be compatible with original equipment manufacturers' tips, ensuring complete compatibility and a consistent fit. One of the widest product offerings combined with our engineering expertise stringent QA/QC enables you to get optimum performance no matter which automated liquid handler you are working with- Compatible with Beckman Coulter™ Biomek™ FX, Biomek NX & Multimek™, Biomek 3000
- Certified DNase-/RNase-free, nonpyrogenic
- With or without aerosol-resistant filters
- Extra long tip
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
30 μL | |
Sterile | |
DNase, RNase Free | |
Beckman Coulter Biomek FX, Beckman Coulter Biomek NX, Beckman Coulter Biomek 3000 |
Filtered | |
Clear | |
Extended Length, Low Retention | |
Polypropylene |