Learn More
Enzo Life Sciences Axin-2 (human), (recombinant) (20µg)
SDP
Supplier:  Enzo Life Sciences ENZPRT1310020
Encompass_Preferred
Alternative name: Axis inhibition protein 2, Axin-like protein, Axil, Conductin. Concentration: 0.05 µg/µl. Purity: ≥70%. Formulation: Liquid. In 50mM TRIS-HCl, pH 7.5, containing 50mM NaCl, 10mM gluthathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. Source: Produced in Sf9 insect cells. Produced in a baculovirus expression system. Long Term Storage: -80°C.
Catalog No. 50-201-3923
Item Discontinued This item has been discontinued by Fisher Scientific. Please to view product availability in your area.