Learn More
Perkin Elmer US LLC Autosampler Hangdown Wire Kit - Qty. 5
Supplier: Perkin Elmer US LLC N5370488
Hangdown wires suspend down from the TGA Furnace Balance Mechanism and catch a sample pan's stirrup and lift the pan from the Autosampler and into the furnace. The hangdown wires come in a variety of materials and kit sizes.