Learn More
Chemglass Life Sciences Autoradiography Film, 8" x 10"
Supplier: Chemglass Life Sciences CLS190602
Autoradiography Film, 8" x 10". • Ideal for Chemiluminescence• Suitable for use with 14C, 32P, 33P, 125I, 35S• Compatible with Automatic Processors• Packaged in Lined, Light-Safe, Dispenser Boxes of 100 Sheets per Box• High Contrast• Reduced Background• Publication Quality• Double EmulsionBluBlot™ HS is a high quality, blue sensitive film that is ideal for a wide range of electrophoretic and autoradiography applications. It can be used with calcium tungstate or blue-emitting rare-earth screens. BluBlot™ HS exhibits excellent sensitivity and contrast characteristics and is coated on both sides for improved consistency and overall performance. It reliably produces sharp, bright images that are easily visualized and suitable for publication.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.