Learn More
Azzota Corp Automatic Polarimeter, Angle Of Rotation Range -45Deg. To +45Deg., Resolution 0.005Deg., Sodium Lamp, Accuracy +/- 0.010Deg., Repeatability 0.010Deg., Speed 1.30Deg./Second, Tube Length Up To 200 mm
Supplier: Azzota Corp AP45
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Automatic Polarimeter, Angle of Rotation Range -45deg. to +45deg., Resolution 0.005deg., Sodium lamp, Accuracy +/- 0.010deg., Repeatability 0.010deg., Speed 1.30deg./second, Tube length up to 200 mm