Learn More
Electron Microscopy Sciences Pipette Jar, Replacement for Automatic Pipette Washing Set
Supplier: Electron Microscopy Sciences 6437715
Solve the problem of washing pipettes quickly, safely and with little effort. To use allow dirty pipettes to soak in the pipette basket in the pipette jar and when ready the basket is transferred to the washer where a constant flow of tap water rinses the pipettes clean.