Learn More
Fujifilm Healthcare Americas Corp Autokit Glucose
Supplier:  Fujifilm Healthcare Americas Corp 99703001
Encompass_Preferred
The Autokit Glucose kit is an in vitro assay utilizing an enzymatic method for the quantitative determination of glucose in serum, plasma, and urine. Package size: Kit includes: Buffer Solution- 2 x 150 mL, Color Reagent (When reconstituted)- 2 x for 150 mL, Standard Solution I- 1 x 10 mL and Standard Solution II- 1 x 10 mL bottles. This product is for Research Use Only, not for use in diagnostic procedures. Contact supplier for additional information at www.ivd.fujimed.com.
Catalog No. NC9927772
Explore available promotions