Learn More
Perkin Elmer US LLC Auto-Ignite FID with PPC Add-On Kit, 230 V
Supplier: Perkin Elmer US LLC N6120168
Kit includes: detector assembly with heater and sensor, heater block, igniter, programmable pneumatics, and controls for detector combustion gases. Requires, but does not include, amplifier (N6120162) and AutoSystem XL firmware revision 3.3 or higher. The AutoSystem XL must be PPC ready. If not, a PPC upgrade kit (N6120146) is required.