Learn More
Perkin Elmer US LLC Auto-Ignite FID with PPC, 120 V
Supplier:  Perkin Elmer US LLC N6120167
Encompass_Preferred
Request Bulk Order
Kit includes: detector assembly with heater and sensor, heater block, igniter, programmable pneumatics, and controls for detector combustion gases. Requires, but does not include, amplifier (N6120162) and AutoSystem XL firmware revision 3.3 or higher. The AutoSystem XL must be PPC ready. If not, a PPC upgrade kit (N6120146) is required.
Catalog No. 50-211-3745