Learn More
Perkin Elmer US LLC Auto-Ignite FID Add-On Kit - 120 V Clarus, Manual Pneumatics
Supplier: Perkin Elmer US LLC N6520018
Kit includes: detector assembly with heater and sensor, heater block, igniter, hydrogen pressure regulator, and needle valve. Requires AutoSystem XL to have firmware revision 3.3 or higher.