Learn More
Electron Microscopy Sciences Aurion Blocking Solution Goat 30 ML
Supplier: Electron Microscopy Sciences 25596
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Aurion has developed Blocking Solutions which effectively block ‘sticky' surfaces by multi-point hydrophobic and charge-based interactions. The Blocking Solutions are specifically designed to meet the characteristics of each type of secondary conjugate. With adequate blocking and incubation media, background reactions are minimized whereas specific reactions are not hampered.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.