Learn More
Mystaire™ Aura™ Elite Series Ductless Fume Hood Accessory, HEPA Filters
SDP
For Aura Elite Series 30, 42 and 54 in. Ductless Fume Hoods. Provides operator safety from potentially harmful airborne particulate.
Supplier:  Mystaire™ MY2820
 View more versions of this product
Catalog No. 15-338-454
Add to Cart
Description
Description
  • Provides 99.97% efficient down to 0.3μ particles
Specifications
Specifications
HEPA
Aura Elite 42 Ductless Fume Hood
Product Suggestions
Product Suggestions