Learn More
Mystaire™ Aura™ Elite Series Ductless Fume Hood Accessory, HEPA Filters
For Aura Elite Series 30, 42 and 54 in. Ductless Fume Hoods. Provides operator safety from potentially harmful airborne particulate.
Supplier: Mystaire™ MY3820
Description
- Provides 99.97% efficient down to 0.3μ particles
Specifications
HEPA | |
Aura Elite 30 and 54 Ductless Fume Hood |