Learn More
Perkin Elmer US LLC ATR Crystal Kit - Germanium
Supplier: Perkin Elmer US LLC L1860317
This kit is a recommended upgrade option for the Spotlight 400 and 400 N. It allows the imaging of samples up to 10 mm thick. The larger crystal enables 400 µm areas to be collected to get more information from the sample quickly.