Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHOWA™ 317 Hi-Vis Latex Coated Seamless Knit Polyester Gloves
Click to view available options
Size:
Small
Medium
Large
X-Large
Description
- Hardwearing gloves designed to provide protection in low-light, ideal for use indoors and out
- Textured latex coating on the palms protects hands and enhances grip
- Elastic wrist cuff keeps dirt and debris out of the gloves
Specifications
Construction | Seamless Knit |
Coating Material | Latex |
Color | Hi-Vis Yellow, Orange |
Exterior Finish | Fully Textured |
Hand | Specific |
Material | Polyester |
Product Type | Seamless Knit Gloves |
Size | Small |
EN388 Standard | 2142X |
For Use With (Application) | Heavy Equipment Handling, Maintenance, and Shipping/Receiving |
Show More |