Learn More
Thermo Scientific ATC-3 ANION TRAP COLUMN
Supplier: Thermo Scientific 059660
Anion Trap Column(ATC)-3; 4mm; 9 x 24mm; Bead dia.: 55um; Capacity: 1.5 meq; High-capacity, low-efficiency, anion-exchange resin in the hydroxide form