Learn More
Across International 250C UL Certified 0.9 CF Vacuum Oven 5 Sided Heat - 110V 60Hz
Supplier: Across International AT09UL110
AT09 series digital desktop vacuum ovens feature a 0.9 cubic foot easy-to-clean stainless steel chamber with a large dual-layer tempered glass safety window and small footprint. These ovens can greatly reduce drying time by maintaining a consistent vacuum level within the chamber. Introducing the new Ai UL certified 0.9 cubic foot vacuum ovens, this certification is required by many municipalities, and it also provides a higher standard of equipment safety and lifespan.Also come standard are the secondary over-temp protection dial, all stainless steel tubing/compression fittings, stainless steel vacuum/vent valve, oil-filled vacuum gauge, 5 sided pad heating technology and two year warranty. The thermal-conductive aluminum pan shelves provide excellent temperature uniformity inside the chamber, while our 4th gen LCD low proportional gain temperature controllers keep your oven temperature accurate.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.