Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ASTM™ Chlordane (Mixture of isomers), 1,000 μg/mL in Isooctane, MilliporeSigma™ Supelco™
Supplier: MilliporeSigma Supelco 48065U
Description
Agriculture, Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal CareSpecifications
Ampule | |
57-74-9 | |
ASTM | |
1,000 μg/mL in isooctane |
1 mL | |
Food and Cosmetic Component Standard | |
TraceCERT™ Analytical Standard | |
-12°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
ASTM is a registered trademark of American Society for Testing and Materials
Spot an opportunity for improvement?
Provide Content Correction